CH4102153512 ChemBridge Corporation

4-[(isobutyrylamino)methyl]benzoic acid

  • MDL number MFCD08593372
  • Cas Number : 908495-37-4
  • Molecular Weight : 221.2524

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071245

Safety Information

MSDS https://www.hit2lead.com/sds/9071245_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart