CH4102153520 ChemBridge Corporation

2-[(5-amino-1,3,4-thiadiazol-2-yl)thio]-N-cyclopropylacetamide

  • MDL number MFCD03609314
  • Cas Number : 876532-11-5
  • Molecular Weight : 230.3105

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7203643

Safety Information

MSDS https://www.hit2lead.com/sds/7203643_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart