CH4102153525 ChemBridge Corporation

2-amino-N-(2-thienylmethyl)benzamide

  • MDL number MFCD00116353
  • Cas Number : 218158-03-3
  • Molecular Weight : 232.3015

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071281

Safety Information

MSDS https://www.hit2lead.com/sds/9071281_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart