CH4102153532 ChemBridge Corporation

2-chloro-N-[5-(tetrahydro-2-furanyl)-1,3,4-thiadiazol-2-yl]acetamide

  • MDL number MFCD08143046
  • Cas Number : 887345-06-4
  • Molecular Weight : 247.7019

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071298

Safety Information

MSDS https://www.hit2lead.com/sds/9071298_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart