CH4102153547 ChemBridge Corporation

1-(2-chloroethoxy)-4-methoxybenzene

  • MDL number MFCD00859185
  • Cas Number : 3383-74-2
  • Molecular Weight : 186.6354

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071331

Safety Information

MSDS https://www.hit2lead.com/sds/9071331_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart