CH4102153549 ChemBridge Corporation

1-(4-hydroxyphenyl)-2-(methylamino)ethanone

  • MDL number MFCD01673131
  • Cas Number : 21213-89-8
  • Molecular Weight : 165.1891

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071337

Safety Information

signalword Warning
hcodes 6.1
MSDS https://www.hit2lead.com/sds/9071337_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart