CH4102153555 ChemBridge Corporation

4-(diethylamino)-2-methylbenzaldehyde

  • MDL number MFCD06800551
  • Cas Number : 92-14-8
  • Molecular Weight : 191.2695

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071344

Safety Information

signalword Warning
hcodes 6.1
MSDS https://www.hit2lead.com/sds/9071344_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart