CH4102153558 ChemBridge Corporation

3-[(3,3-dimethylbutanoyl)amino]-4-methylbenzoic acid

  • MDL number MFCD08569895
  • Cas Number : 915921-64-1
  • Molecular Weight : 249.3056

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071354

Safety Information

MSDS https://www.hit2lead.com/sds/9071354_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart