CH4102153560 ChemBridge Corporation

N-(2-furylmethyl)-2-propen-1-amine

  • MDL number MFCD07410484
  • Cas Number : 53175-33-0
  • Molecular Weight : 137.179

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071357

Safety Information

MSDS https://www.hit2lead.com/sds/9071357_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart