CH4102153579 ChemBridge Corporation

2-methyl-3-[(tetrahydro-2-furanylcarbonyl)amino]benzoic acid

  • MDL number MFCD06091787
  • Cas Number : 876890-63-0
  • Molecular Weight : 249.2625

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071423

Safety Information

MSDS https://www.hit2lead.com/sds/9071423_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart