CH4102153584 ChemBridge Corporation

2-{[(3-methyl-2-thienyl)methyl]amino}ethanol

  • MDL number MFCD04594770
  • Cas Number : 869941-84-4
  • Molecular Weight : 171.2599

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071449

Safety Information

MSDS https://www.hit2lead.com/sds/9071449_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart