CH4102153592 ChemBridge Corporation

3-(2,5-dimethylphenoxy)-N-methyl-1-propanamine

  • MDL number MFCD08691955
  • Cas Number : 915924-19-5
  • Molecular Weight : 193.2854

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071476

Safety Information

MSDS https://www.hit2lead.com/sds/9071476_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart