CH4102153598 ChemBridge Corporation

5-bromo-N-methyl-2-thiophenecarboxamide

  • MDL number MFCD08691957
  • Cas Number : 98453-26-0
  • Molecular Weight : 220.0869

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071495

Safety Information

MSDS https://www.hit2lead.com/sds/9071495_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart