CH4102153601 ChemBridge Corporation

1-(chloroacetyl)-6-fluoro-2-methyl-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD03383529
  • Cas Number : 544692-44-6
  • Molecular Weight : 241.6891

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7236294

Safety Information

MSDS https://www.hit2lead.com/sds/7236294_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart