CH4102153616 ChemBridge Corporation

2,4-dimethyl-5-(1H-1,2,4-triazol-1-ylmethyl)benzaldehyde

  • MDL number MFCD07162613
  • Cas Number : 894365-26-5
  • Molecular Weight : 215.2511

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071543

Safety Information

MSDS https://www.hit2lead.com/sds/9071543_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart