CH4102153625 ChemBridge Corporation

3-(4-fluorophenoxy)-N-methyl-1-propanamine

  • MDL number MFCD06800761
  • Cas Number : 883542-69-6
  • Molecular Weight : 183.2227

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071556

Safety Information

MSDS https://www.hit2lead.com/building-blocks/9071556
Availability details for
Quantity
  • Availability
  • Add to Cart