CH4102153636 ChemBridge Corporation

N-(5-bromo-2-pyridinyl)-2-chloroacetamide

  • MDL number MFCD00461174
  • Cas Number : 141454-61-7
  • Molecular Weight : 249.4923

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071591

Safety Information

MSDS https://www.hit2lead.com/sds/9071591_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart