CH4102153641 ChemBridge Corporation

2,4-dimethyl-5-[(4-methyl-1-piperidinyl)methyl]benzaldehyde

  • MDL number MFCD07156278
  • Cas Number : 894207-03-5
  • Molecular Weight : 245.3599

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071600

Safety Information

MSDS https://www.hit2lead.com/sds/9071600_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart