CH4102153643 ChemBridge Corporation

3-chloro-N-mesitylpropanamide

  • MDL number MFCD03385087
  • Cas Number : 100141-43-3
  • Molecular Weight : 225.7145

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7246527

Safety Information

MSDS https://www.hit2lead.com/sds/7246527_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart