CH4102153645 ChemBridge Corporation

3-chloro-N-(2-thienylmethyl)propanamide

  • MDL number MFCD03394727
  • Cas Number : 21403-26-9
  • Molecular Weight : 203.6891

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7248309

Safety Information

MSDS https://www.hit2lead.com/building-blocks/7248309
Availability details for
Quantity
  • Availability
  • Add to Cart