CH4102153653 ChemBridge Corporation

2,5-dimethyl-3-[(2-methyl-1-piperidinyl)methyl]benzaldehyde

  • MDL number MFCD07156457
  • Cas Number : 894213-68-4
  • Molecular Weight : 245.3599

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071637

Safety Information

MSDS https://www.hit2lead.com/sds/9071637_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart