CH4102153672 ChemBridge Corporation

N-(2-pyridinylmethyl)-2-propen-1-amine

  • MDL number MFCD07411573
  • Cas Number : 62402-16-8
  • Molecular Weight : 148.205

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071688

Safety Information

MSDS https://www.hit2lead.com/sds/9071688_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart