CH4102153673 ChemBridge Corporation

N-(4-sec-butylphenyl)-2-chloroacetamide

  • MDL number MFCD03147362
  • Cas Number : 20331-26-4
  • Molecular Weight : 225.7145

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7258079

Safety Information

MSDS https://www.hit2lead.com/sds/7258079_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart