CH4102153677 ChemBridge Corporation

2-[(tert-butylamino)carbonyl]cyclohexanecarboxylic acid

  • MDL number MFCD07700900
  • Cas Number : 69049-86-1
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7259223

Safety Information

MSDS https://www.hit2lead.com/sds/7259223_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart