CH4102153693 ChemBridge Corporation

4-[(2-isopropyl-6-methylphenyl)amino]-4-oxobutanoic acid

  • MDL number MFCD06065527
  • Cas Number : 904797-70-2
  • Molecular Weight : 249.3056

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071748

Safety Information

MSDS https://www.hit2lead.com/sds/9071748_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart