CH4102153704 ChemBridge Corporation

3-chloro-N-cyclooctylpropanamide

  • MDL number MFCD03375171
  • Cas Number : 543711-67-7
  • Molecular Weight : 217.7356

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7276810

Safety Information

MSDS https://www.hit2lead.com/sds/7276810_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart