CH4102153738 ChemBridge Corporation

5-bromo-N-cyclopropyl-2-furamide

  • MDL number MFCD01354260
  • Cas Number : 352682-00-9
  • Molecular Weight : 230.0586

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7284172

Safety Information

MSDS https://www.hit2lead.com/sds/7284172_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart