CH4102153761 ChemBridge Corporation

5-bromo-N-propyl-2-thiophenecarboxamide

  • MDL number MFCD08593343
  • Cas Number : 908494-85-9
  • Molecular Weight : 248.1401

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071929

Safety Information

MSDS https://www.hit2lead.com/sds/9071929_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart