CH4102153773 ChemBridge Corporation

3-[(2-methyl-2-propen-1-yl)oxy]benzaldehyde

  • MDL number MFCD07392462
  • Cas Number : 38002-95-8
  • Molecular Weight : 176.2118

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071964

Safety Information

MSDS https://www.hit2lead.com/sds/9071964_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart