CH4102153781 ChemBridge Corporation

4-(acetylamino)-3-methylbenzoic acid

  • MDL number MFCD08691999
  • Cas Number : 37901-92-1
  • Molecular Weight : 193.1992

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071980

Safety Information

MSDS https://www.hit2lead.com/building-blocks/9071980
Availability details for
Quantity
  • Availability
  • Add to Cart