CH4102153783 ChemBridge Corporation

N-(2-aminoethyl)-2-chlorobenzamide

  • MDL number MFCD04371105
  • Cas Number : 65389-77-7
  • Molecular Weight : 198.6494

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071983

Safety Information

MSDS https://www.hit2lead.com/sds/9071983_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart