CH4102153795 ChemBridge Corporation

5-bromo-N-isopropyl-2-thiophenecarboxamide

  • MDL number MFCD08593344
  • Cas Number : 908494-87-1
  • Molecular Weight : 248.1401

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9072032

Safety Information

MSDS https://www.hit2lead.com/building-blocks/9072032
Availability details for
Quantity
  • Availability
  • Add to Cart