CH4102153810 ChemBridge Corporation

(3-butyryl-1H-indol-1-yl)acetic acid

  • MDL number MFCD08676634
  • Cas Number : 892676-98-1
  • Molecular Weight : 245.2738

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9072054

Safety Information

MSDS https://www.hit2lead.com/sds/9072054_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart