CH4117773736 ChemBridge Corporation

[3-(4-bromophenoxy)propyl]methylamine oxalate

  • MDL number MFCD08691784
  • Cas Number : 915922-44-0
  • Molecular Weight : 244.1283

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7710120

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/7710120_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart