CH4118669799 ChemBridge Corporation

[2-(2-bromophenoxy)ethyl]ethylamine hydrochloride

  • MDL number MFCD08691845
  • Cas Number : 915920-58-0
  • Molecular Weight : 244.1283

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9023838

Safety Information

MSDS https://www.hit2lead.com/sds/9023838_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart