CH4120981011 ChemBridge Corporation

[2-isopropoxy-5-(trifluoromethyl)phenyl]amine hydrochloride

  • MDL number MFCD00616961
  • Cas Number : 121307-23-1
  • Molecular Weight : 219.2036

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6369011

Safety Information

MSDS https://www.hit2lead.com/sds/6369011_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart