CH4120981083 ChemBridge Corporation

N-{[3-(4-methylphenyl)-1,2,4-oxadiazol-5-yl]methyl}ethanamine hydrochloride

  • MDL number MFCD07087663
  • Cas Number : 890324-84-2
  • Molecular Weight : 217.267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9026378

Safety Information

MSDS https://www.hit2lead.com/sds/9026378_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart