CH4120981086 ChemBridge Corporation

N-{[3-(3-methylphenyl)-1,2,4-oxadiazol-5-yl]methyl}ethanamine hydrochloride

  • MDL number MFCD06752199
  • Cas Number : 890324-23-9
  • Molecular Weight : 217.267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9032218

Safety Information

MSDS https://www.hit2lead.com/sds/9032218_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart