CH4120981089 ChemBridge Corporation

cyclopentyl 4-aminobenzoate hydrochloride

  • MDL number MFCD08070526
  • Cas Number : 37005-75-7
  • Molecular Weight : 205.253

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9038244

Safety Information

MSDS https://www.hit2lead.com/sds/9038244_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart