CH4151861423 ChemBridge Corporation

N-propylcyclohexanamine

  • MDL number MFCD03210942
  • Cas Number : 3592-81-2
  • Molecular Weight : 141.2539

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6591639

Safety Information

MSDS https://www.hit2lead.com/building-blocks/6591639
Availability details for
Quantity
  • Availability
  • Add to Cart