CH4151861435 ChemBridge Corporation

N-methyl-1-phenylpropan-1-amine

  • MDL number MFCD04038424
  • Cas Number : 7713-71-5
  • Molecular Weight : 149.2328

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6871471

Safety Information

MSDS https://www.hit2lead.com/sds/6871471_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart