CH4182074713 ChemBridge Corporation

5-isobutyl-1H-pyrazole-3-carbaldehyde

  • MDL number MFCD09864235
  • Cas Number : 1025032-18-1
  • Molecular Weight : 152.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003141

Safety Information

MSDS https://www.hit2lead.com/sds/4003141_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart