CH4182074799 ChemBridge Corporation

3-(chloromethyl)-5-isopropyl-1,2,4-oxadiazole

  • MDL number MFCD09864313
  • Cas Number : 189130-87-8
  • Molecular Weight : 160.6015

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4013193

Safety Information

MSDS https://www.hit2lead.com/sds/4013193_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart