CH4182074822 ChemBridge Corporation

[4-(1-morpholin-4-ylethyl)phenyl]boronic acid

  • MDL number MFCD09864341
  • Cas Number : 1226814-83-0
  • Molecular Weight : 235.0872

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4015557

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4015557_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart