CH4182074824 ChemBridge Corporation

rac-(3S,4R)-3-benzyl-1-methylpiperidin-4-amine

  • MDL number MFCD09802316
  • Cas Number : 952480-26-1
  • Molecular Weight : 204.3113

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4015684

Safety Information

MSDS https://www.hit2lead.com/sds/4015684_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart