CH4182074886 ChemBridge Corporation

2-methoxybutanoic acid

  • MDL number MFCD09864397
  • Cas Number : 56674-69-2
  • Molecular Weight : 118.1311

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4020880

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4020880
Availability details for
Quantity
  • Availability
  • Add to Cart