CH4182074907 ChemBridge Corporation

1-(5-methyl-1H-benzimidazol-2-yl)propan-1-amine

  • MDL number MFCD09055212
  • Cas Number : 884504-85-2
  • Molecular Weight : 189.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003325

Safety Information

MSDS https://www.hit2lead.com/sds/4003325_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart