CH4182074959 ChemBridge Corporation

N-benzyl-1-cyclopropylmethanamine

  • MDL number MFCD03129358
  • Cas Number : 116373-23-0
  • Molecular Weight : 161.2435

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4001187

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4001187
Availability details for
Quantity
  • Availability
  • Add to Cart