CH4182075015 ChemBridge Corporation

N-methyl-3-piperidin-1-ylpropan-1-amine

  • MDL number MFCD07186366
  • Cas Number : 86010-41-5
  • Molecular Weight : 156.2685

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4004398

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4004398
Availability details for
Quantity
  • Availability
  • Add to Cart