CH4182075058 ChemBridge Corporation

1-ethyl-1H-imidazole-2-carbaldehyde

  • MDL number MFCD07380875
  • Cas Number : 111851-98-0
  • Molecular Weight : 124.1405

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4004954

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4004954
Availability details for
Quantity
  • Availability
  • Add to Cart